Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MAP1B/MAP5 (Microtubule-Associated Protein) Ab-1, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ MAP1B/MAP5 (Microtubule-Associated Protein) Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS248P1ABX

 View more versions of this product

Catalog No. MS248P1ABX

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Rat
Antigen MAP1B/MAP5 (Microtubule-Associated Protein) Ab-1
Host Species Mouse
Isotype IgG1
Regulatory Status RUO
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Quantity 100µL
Format Concentrated
Clone 3G5
Immunogen Purified bovine brain MAPs
Formulation Ab without BSA/Azide (Frozen)
Research Discipline Cancer and Tumor Biology
Type Primary
Classification Monoclonal
Concentration 1mg/mL
Conjugate Unlabeled
Item Description 100uL; 1mg/mL; Unlabeled; Purified without BSA and azide

Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ MAP1B/MAP5 (Microtubule-Associated Protein) Ab-1, Mouse Monoclonal Antibody.

Microtubule-associated protein 1B (MAP1B, also known as MAP5, MAP1.2, MAP1(x), or MAP1X) is different from MAP1A (also known as MAP1 or MAP1.1). Reportedly, MAP1B/MAP5 is an early MAP; it is present at high levels in embryonic and newborn rat brain and declines several-fold upon brain maturation. In several cellular situations, MAP1B/MAP5 is the first neuronal MAP to appear, and it is found in neurites from their very first emergence from the cell body. Expression of MAP1B/MAP5 is induced by nerve growth factor.

Host Species: Mouse

Clone: 3G5

Isotype: IgG1

Species Reactivity: Human, Cow and Rat. Does not react with chicken. Others not known.

Epitope: Not determined

Immunogen: Purified bovine brain MAPs

Molecular Weight: 320kDa doublet

Positive Control: Brain

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not verified)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO