missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

American Educational Products Map Reading Kit

Teaches basic map reading skills.

Manufacturer:  American Educational Products HUB625

 View more versions of this product

Catalog No. S45066



Includes: Plastic terrain model with five corresponding stacking contour sheets, five unprinted stacking contour sheets for projects, wall map of the relief model with 50 student project maps, grease pencil and study guide

Description

Description

  • Learn how to determine location and elevation and understand scales and symbols
  • Plastic can be marked and erased
  • L × W × H: 10 × 2 × 10 in. (25.4 × 5.1 × 25.4cm)

Specifications

Specifications

Map Reading Kit
Teaches basic map reading skills
10 in.
Plastic terrain model with five corresponding stacking contour sheets, five unprinted stacking contour sheets for projects, wall map of the relief model with 50 student project maps, grease pencil and study guide
25.4cm
2 in.
10 x 2 x 10 in. (25.4 x 5.1 x 25.4cm)
Education
6 to 12
25.4cm
10 in.
Each
5.1cm
Documents