missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

American Educational Products Map Reading Kit

Teaches basic map reading skills.

$30.34 - $182.06

Specifications

Dimensions (L x W x H) 10 x 2 x 10 in. (25.4 x 5.1 x 25.4cm)
For Use With (Application) Teaches basic map reading skills
Grade 6 to 12
Includes Plastic terrain model with five corresponding stacking contour sheets, five unprinted stacking contour sheets for projects, wall map of the relief model with 50 student project maps, grease pencil and study guide
View More Specs

Includes: Plastic terrain model with five corresponding stacking contour sheets, five unprinted stacking contour sheets for projects, wall map of the relief model with 50 student project maps, grease pencil and study guide


Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Quantity Price Quantity  
Catalog Number Mfr. No. Product Type Quantity Price Quantity  
S45066 American Educational Products
HUB625
Map Reading Kit Each Each for $182.06
S45066A American Educational Products
HUB627
Refill Student Project Maps 50 Pack of 50 Each for $30.34
Specifications

Description

  • Learn how to determine location and elevation and understand scales and symbols
  • Plastic can be marked and erased
  • L × W × H: 10 × 2 × 10 in. (25.4 × 5.1 × 25.4cm)

Description & Specifications

Specifications

10 x 2 x 10 in. (25.4 x 5.1 x 25.4cm)
Teaches basic map reading skills
6 to 12
Plastic terrain model with five corresponding stacking contour sheets, five unprinted stacking contour sheets for projects, wall map of the relief model with 50 student project maps, grease pencil and study guide