Please login to your online account to display your discounted pricing

Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers


Achieve secure, repeatable crimps or remove seals with minimized risk of damage.

Manufacturer: thermo scientific™  C4030100

 View more versions of this product

 Disclaimers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Catalog No. 03-375-52A

  • / Each
 

Description & Specifications

Specifications

Product Line National
For Use With 30mm crimp seals
Description Crimper, For 32mm crimp seals
Type Manual Crimper
Item Description C4030-100
Vial Closure Size 30mm
Vial Brand National

Achieve secure, repeatable crimps or remove seals with minimized risk of damage when you use Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers, also for use with National or Chromacol vials.

Lightweight, precision crimpers attach seals to chromatography or serum vials.

  • Factory set to propery crimp standard-thickness seals
  • Adjustable handle setting scree allows proper crimps with septa of varying thickness