Please login to your online account to display your discounted pricing

Alfa Aesar™ Manoalide, 98%

Chemical Identifiers

Chemical Name or Material Manoalide
CAS 75088-80-1
Formula Weight 416.56
MDL Number MFCD00153826
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67341MCR

 
alfa aesar™
J67341MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 416.56
Chemical Name or Material Manoalide
Assay 98%
MDL Number MFCD00153826
Physical Form Solid
CAS 75088-80-1
Storage Note 1 -30° to -10°C
Color Colorless to off-white
Quantity 1mg
Safety and Handling

-30° to -10°C