Please login to your online account to display your discounted pricing

Mannitol, ACS, EMD Millipore

$760.91 - $760.91

Chemical Identifiers

Chemical Name or Material Mannitol
Name Note For The Determination of Boric Acid
CAS 69-65-8
Physical Form Powder
Molecular Formula C6H14O6
Formula Weight 182.17
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MMX02141

 
emd millipore corporation
MX02141
3kg Each for $760.91
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 182.17
Color White
Quantity 3kg
Bulk Density 400 to 500kg/m3
Packaging Poly Pail
Boiling Point 290 to 295°C
Melting Point 164° to 169°C
Density 1.49g/cm3 (20°C)
Chemical Name or Material Mannitol
Name Note For The Determination of Boric Acid
Ignition Point 410°C
Molecular Formula C6H14O6
CAS 69-65-8
Recommended Storage Store without restrictions
Safety and Handling

Store without restrictions