missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Manidipine Dihydrochloride, TCI America™

$221.53 - $768.48

Chemical Identifiers

CAS 89226-75-5
Molecular Formula C35H40Cl2N4O6
Molecular Weight (g/mol) 683.627
MDL Number MFCD00896434
InChI Key JINNGBXKBDUGQT-UHFFFAOYSA-N
Synonym manidipine hydrochloride
PubChem CID 150762
IUPAC Name 5-O-[2-(4-benzhydrylpiperazin-1-yl)ethyl] 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;dihydrochloride
SMILES CC1=C(C(C(=C(N1)C)C(=O)OCCN2CCN(CC2)C(C3=CC=CC=C3)C4=CC=CC=C4)C5=CC(=CC=C5)[N+](=O)[O-])C(=O)OC.Cl.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M22251G View Documents TCI America
M22251G
1g Each for $221.53
 
M22255G View Documents TCI America
M22255G
5g Each for $768.48
 
Specifications

Chemical Identifiers

89226-75-5
683.627
JINNGBXKBDUGQT-UHFFFAOYSA-N
150762
CC1=C(C(C(=C(N1)C)C(=O)OCCN2CCN(CC2)C(C3=CC=CC=C3)C4=CC=CC=C4)C5=CC(=CC=C5)[N+](=O)[O-])C(=O)OC.Cl.Cl
C35H40Cl2N4O6
MFCD00896434
manidipine hydrochloride
5-O-[2-(4-benzhydrylpiperazin-1-yl)ethyl] 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;dihydrochloride

Specifications

White-Yellow
1g
C35H40Cl2N4O6
2811
JINNGBXKBDUGQT-UHFFFAOYSA-N
5-O-[2-(4-benzhydrylpiperazin-1-yl)ethyl] 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;dihydrochloride
150762
Crystal-Powder at 20°C
Manidipine Dihydrochloride
211°C
89226-75-5
MFCD00896434
manidipine hydrochloride
CC1=C(C(C(=C(N1)C)C(=O)OCCN2CCN(CC2)C(C3=CC=CC=C3)C4=CC=CC=C4)C5=CC(=CC=C5)[N+](=O)[O-])C(=O)OC.Cl.Cl
683.627
683.63
≥98.0% (HPLC,T)
Safety and Handling

Safety and Handling

RTECSNumber : US7975300

TSCA : No