missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Manidipine Dihydrochloride 98.0+%, TCI America™

Manufacturer:  Tci America M22255G

 View more versions of this product

Catalog No. M22255G



Specifications

Specifications

Manidipine Dihydrochloride
211°C
89226-75-5
MFCD00896434
manidipine hydrochloride
CC1=C(C(C(=C(N1)C)C(=O)OCCN2CCN(CC2)C(C3=CC=CC=C3)C4=CC=CC=C4)C5=CC(=CC=C5)[N+](=O)[O-])C(=O)OC.Cl.Cl
683.627
683.63
≥98.0% (HPLC,T)
White-Yellow
5g
C35H40Cl2N4O6
2811
JINNGBXKBDUGQT-UHFFFAOYSA-N
5-O-[2-(4-benzhydrylpiperazin-1-yl)ethyl] 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;dihydrochloride
150762
Crystal-Powder at 20°C

Chemical Identifiers

89226-75-5
683.627
JINNGBXKBDUGQT-UHFFFAOYSA-N
150762
CC1=C(C(C(=C(N1)C)C(=O)OCCN2CCN(CC2)C(C3=CC=CC=C3)C4=CC=CC=C4)C5=CC(=CC=C5)[N+](=O)[O-])C(=O)OC.Cl.Cl
C35H40Cl2N4O6
MFCD00896434
manidipine hydrochloride
5-O-[2-(4-benzhydrylpiperazin-1-yl)ethyl] 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;dihydrochloride
Safety and Handling

Safety and Handling

RTECSNumber : US7975300

TSCA : No

Documents
Promotions

Promotions