Please login to your online account to display your discounted pricing

Manganous Sulfate, Monohydrate, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$392.73 - $1,321.96

Chemical Identifiers

Chemical Name or Material Manganous Sulfate
CAS 10034-96-5
Name Note Monohydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula MnSO4·H2O
Formula Weight 169.02
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003377

 
avantor performance
255005
Poly Bottle 2.5kg Each for $1,321.96

02003378

 
avantor performance
255001
Poly Bottle 500g Each for $392.73
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 169.02
Chemical Name or Material Manganous Sulfate
CAS 10034-96-5
Name Note Monohydrate
Packaging Poly Bottle
Physical Form Powder
Specific Gravity 1L = 2.95kg
Molecular Formula MnSO4·H2O
Quantity 2.5kg