Please login to your online account to display your discounted pricing

Manganous Sulfate, Monohydrate, ACS Grade, 98.0 to 101.0%, LabChem™

$209.94 - $209.94

Chemical Identifiers

Chemical Name or Material Manganous Sulfate
CAS 10034-96-5
CAS Max % 100
Name Note Monohydrate
Purity Grade ACS Grade
Linear Formula MnSO4·H2O
Formula Weight 169.01
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC165551

 
lab chem
LC165551
500g Each for $209.94
Description & Specifications

Specifications

Formula Weight 169.01
Chemical Name or Material Manganous Sulfate
Identification Passes Test
Name Note Monohydrate
Packaging Poly Bottle
Purity Grade ACS Grade
Physical Form Powder
Reducing Substances Passes Test
Assay 98.0 to 101.0%
Solubility Soluble in water
Storage Information Room Temperature
Linear Formula MnSO4·H2O
CAS 10034-96-5
Decomposition Information Sulfur compounds
Boiling Point 850°C
Color Light pink
Melting Point 700°C
Quantity 500g
Health Hazard 1 Warning
CAS Max % 100
Health Hazard 2 GHS H Statement
May cause damage to organs (central nervous system) through prolonged or repeated exposure (inhalation).
Toxic to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement
Do not breathe dust.
Avoid release to the environment.
Get medical advice and attention if you feel unwell.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.
Safety and Handling

Warning

GHS P Statement
Do not breathe dust.
Avoid release to the environment.
Get medical advice and attention if you feel unwell.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
May cause damage to organs (central nervous system) through prolonged or repeated exposure (inhalation).
Toxic to aquatic life with long lasting effects.