missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Manganous Sulfate, 0.31% (w/v) Aqueous Solution, Ricca Chemical

Manufacturer:  Ricca Chemical Company RIR461000020F

 View more versions of this product

Catalog No. R461000020F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Manganous Sulfate
97.0,0.54,0.3
dihydrogen monoxide, dihydrogen oxide
Cubitainer
20L
7732-18-5,7664-93-9,10034-96-5
100.0,0.56,0.31
Liquid
Colorless

Chemical Identifiers

7732-18-5
dihydrogen monoxide, dihydrogen oxide
Documents