missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Manganous Sulfate, 0.31% (w/v), Ricca Chemical

Chemical Identifiers

CAS 7732-18-5
Synonym dihydrogen oxide, dihydrogen monoxide
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
4610-16
View Documents Promotion Details
Ricca Chemical Company
461016
500 mL Natural Poly Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
R46100001A
View Documents Promotion Details
Ricca Chemical Company
R46100001A
1 L Natural Poly Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

7732-18-5
dihydrogen oxide, dihydrogen monoxide

Specifications

7732-18-5,7664-93-9,10034-96-5
100.0,0.56,0.31
Natural Poly Bottle
500 mL
Manganous Sulfate
97.0,0.54,0.3
dihydrogen oxide, dihydrogen monoxide
Aqueous Solution
0.31% (w/v)
SDS
missing translation for 'documents'

missing translation for 'documents'