Please login to your online account to display your discounted pricing

Manganous Chloride, 4-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 125g
Packaging Poly Bottle

Manufacturer: avantor performance  254004

 View more versions of this product

Catalog No. 02003374

  • / Each
  • / Case of 12 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Baker Analyzed™ ACS Reagent
Physical Form Crystals
Formula Weight 197.9
Chemical Name or Material Manganous Chloride
Molecular Formula MnCl2·4H2O
CAS 13446-34-9
Name Note 4-Hydrate
Packaging Poly Bottle
Specific Gravity 1L = 2.01kg
Quantity 125g

Specifications

Chemical Name or Material Manganous Chloride
CAS 13446-34-9
Name Note 4-Hydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula MnCl2·4H2O
Formula Weight 197.9