missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Manganese telluride, 99.9% (metals basis), Thermo Scientific Chemicals
$112.40 - $860.74
Chemical Identifiers
CAS | 12032-88-1 |
---|---|
Molecular Formula | MnTe |
Molecular Weight (g/mol) | 182.54 |
MDL Number | MFCD00049487 |
InChI Key | VMINMXIEZOMBRH-UHFFFAOYSA-N |
Synonym | manganese telluride, telluroxomanganese, manganese monotelluride, manganous telluride, manganese ii telluride, manganese 2+ telluride |
PubChem CID | 82828 |
IUPAC Name | tellanylidenemanganese |
SMILES | [Mn]=[Te] |
Description
Manganese telluride is used in scientific research and developments.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12032-88-1 | |
182.54 | |
VMINMXIEZOMBRH-UHFFFAOYSA-N | |
82828 | |
[Mn]=[Te] |
MnTe | |
MFCD00049487 | |
manganese telluride, telluroxomanganese, manganese monotelluride, manganous telluride, manganese ii telluride, manganese 2+ telluride | |
tellanylidenemanganese |
Specifications
Gray | |
Lump | |
MnTe | |
manganese telluride, telluroxomanganese, manganese monotelluride, manganous telluride, manganese ii telluride, manganese 2+ telluride | |
VMINMXIEZOMBRH-UHFFFAOYSA-N | |
tellanylidenemanganese | |
82828 | |
99.9% | |
(metals basis) |
12032-88-1 | |
1 g | |
MFCD00049487 | |
Insoluble aq. soln.. | |
[Mn]=[Te] | |
182.54 | |
182.54 | |
Manganese telluride |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
EINECSNumber : 234-782-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only