Please login to your online account to display your discounted pricing

Manganese Sulfate Monohydrate (Granular Powder/USP/FCC), Fisher Chemical


Quantity 2.5kg
Packaging Poly Bottle

Manufacturer: fisher scientific company  FLM114212

 View more versions of this product

Catalog No. M114-212

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade FCC/USP
Formula Weight 169.02
Boiling Point 850°C
Melting Point 700°C
Quantity 2.5kg
Physical Form Solid
Assay 98.0 to 102.0% (FCC)
Color Light Red
Molecular Formula MnSO4 · H2O
Viscosity [NULL]
Decomposition Information 850°C
pH 3 to 3.5
Density 3.25g/cm³
Chemical Name or Material Manganese Sulfate Monohydrate
Identification Pass Test (FCC)
Lead (Pb) 4mg/kg max. (FCC)
Loss on Ignition 10.0 to 13.0% (USP)
Loss on Heating 10.0 to 13.0% (FCC)
Residual Solvents Meets Requirements (USP)
Selenium (Se) 0.003% max. (FCC)
Substances not ppt by (NH4)2S 0.5% max. (USP)
MDL Number MFCD00149159
CAS 10034-96-5
ChemAlert Storage Symbol Gray
Health Hazard 2 WARNING!
Packaging Poly Bottle
Arsenic (As) 3mg/kg max. (FCC)
Synonym Manganese (IV) Sulfate Monohydrate
Health Hazard 3 Emergency Overview
May be harmful if swallowed or inhaled. May cause eye, skin, and respiratory tract irritation. Danger of serious damage to health by prolonged exposure. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:0
Instability:0

Specifications

Chemical Name or Material Manganese Sulfate Monohydrate
CAS 10034-96-5
Purity Grade FCC/USP
Physical Form Solid
Assay 98.0 to 102.0% (FCC)
Molecular Formula MnSO4 · H2O
Formula Weight 169.02
MDL Number MFCD00149159
Synonym Manganese (IV) Sulfate Monohydrate
Accessories
 

Emergency Overview
May be harmful if swallowed or inhaled. May cause eye, skin, and respiratory tract irritation. Danger of serious damage to health by prolonged exposure. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:0
Instability:0

WARNING!

SDS & Safety Documents