missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Manganese Sulfate Monohydrate (Granular Powder/USP/FCC), Fisher Chemical

Manufacturer:  Thermo Fisher Scientific FLM11412

 View more versions of this product

Catalog No. M114-12


 

Specifications Description & Specifications

Specifications

700°C
FCC/USP
12kg
Solid
850°C
3 to 3.5
Meets Requirements (USP)
10.0 to 13.0% (FCC)
Poly Pail
Pass Test (FCC)
4mg/kg max. (FCC)
0.5% max. (USP)
Manganese (IV) Sulfate Monohydrate
850°C
169.02
98.0 to 102.0% (USP)
MnSO4 · H2O
Light Red
3.25g/cm³
3mg/kg max. (FCC)
0.003% max. (FCC)
Manganese Sulfate Monohydrate
10034-96-5
MFCD00149159
10.0 to 13.0% (USP)

Chemical Identifiers

10034-96-5
MnSO4 · H2O
MFCD00149159
Manganese Sulfate Monohydrate
FCC/USP
Solid
169.02
Manganese (IV) Sulfate Monohydrate
98.0 to 102.0% (USP)

Safety and Handling

SDS & Safety Documents

WARNING!

Emergency Overview
May be harmful if swallowed or inhaled. May cause eye, skin, and respiratory tract irritation. Danger of serious damage to health by prolonged exposure. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:0
Instability:0