Please login to your online account to display your discounted pricing

Manganese Sulfate Monohydrate (Granular Powder/USP/FCC), Fisher Chemical

$337.00 - $3,826.79

Chemical Identifiers

Chemical Name or Material Manganese Sulfate Monohydrate
CAS 10034-96-5
Purity Grade FCC/USP
Physical Form Solid
Assay 98.0 to 102.0% (FCC)
Molecular Formula MnSO4 · H2O
Formula Weight 169.02
MDL Number MFCD00149159
Synonym Manganese (IV) Sulfate Monohydrate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M114-12

  SDS
thermo fisher scientific
FLM11412
Poly Pail 12kg Each for $3,826.79

M114-500

  SDS
fisher scientific company
FLM114500
Poly Bottle 500g Each for $337.00
Case of 6 Each for $2,022.00

M114-212

  SDS
fisher scientific company
FLM114212
Poly Bottle 2.5kg Each for $1,290.57
Description & Specifications

Specifications

Purity Grade FCC/USP
Formula Weight 169.02
Boiling Point 850°C
Melting Point 700°C
Quantity 12kg
Physical Form Solid
Assay 98.0 to 102.0% (USP)
Molecular Formula MnSO4 · H2O
Viscosity [NULL]
Decomposition Information 850°C
Color Light Red
pH 3 to 3.5
Density 3.25g/cm³
Chemical Name or Material Manganese Sulfate Monohydrate
Loss on Heating 10.0 to 13.0% (FCC)
Packaging Poly Pail
Residual Solvents Meets Requirements (USP)
Selenium (Se) 0.003% max. (FCC)
Arsenic (As) 3mg/kg max. (FCC)
Identification Pass Test (FCC)
Lead (Pb) 4mg/kg max. (FCC)
MDL Number MFCD00149159
CAS 10034-96-5
ChemAlert Storage Symbol Gray
Loss on Ignition 10.0 to 13.0% (USP)
Substances not ppt by (NH4)2S 0.5% max. (USP)
Synonym Manganese (IV) Sulfate Monohydrate
Health Hazard 2 WARNING!
Health Hazard 3 Emergency Overview
May be harmful if swallowed or inhaled. May cause eye, skin, and respiratory tract irritation. Danger of serious damage to health by prolonged exposure. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:0
Instability:0
Safety and Handling
 

Emergency Overview
May be harmful if swallowed or inhaled. May cause eye, skin, and respiratory tract irritation. Danger of serious damage to health by prolonged exposure. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:0
Instability:0

WARNING!