missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals Inc Manganese Sulfate Monohydrate, (Crystalline),MP Biomedicals
Supplier: MP Biomedicals Inc 0215533691
Specifications
Manganese Sulfate Monohydrate | |
Crystalline | |
H2MnO5S | |
manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate | |
O.[Mn++].[O-]S([O-])(=O)=O | |
169.01 | |
CHEBI:86364 |
10034-96-5 | |
1 kg | |
MFCD00149159 | |
ISPYRSDWRDQNSW-UHFFFAOYSA-L | |
manganese(2+) hydrate sulfate | |
177577 | |
169 |
Chemical Identifiers
10034-96-5 | |
169.01 | |
ISPYRSDWRDQNSW-UHFFFAOYSA-L | |
177577 | |
manganese(2+) hydrate sulfate |
H2MnO5S | |
MFCD00149159 | |
manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate | |
CHEBI:86364 | |
O.[Mn++].[O-]S([O-])(=O)=O |
Safety and Handling
Recommended Storage : Room Temperature