missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Manganese Standard, 1000mg/L in 2% HNO3 (Assurance Grade), SPEX CertiPrep™

Manufacturer:  Spex Certiprep Llc SPXPLMN22M

 View more versions of this product

Catalog No. PLMN22M


 

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Specifications

Description

Manganese Standard; SPEX CertiPrep; 30mL; Triple-leached LDPE bottle

AnalyteConcentration
Mn1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Description & Specifications

Specifications

Manganese Standard
0.1,2.0,97.9
Assurance Grade
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
30mL
7439-96-5,7697-37-2,7732-18-5
0.1,2.0,97.9
Triple-leached LDPE bottle
2% HNO3
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

Chemical Identifiers

7439-96-5,7697-37-2,7732-18-5
0.1,2.0,97.9
Assurance Grade
0.1,2.0,97.9
Manganese Standard

Safety and Handling

Warning: Causes skin irritation, serious eye irritation Wear protective gloves/eye/face protection. If in eyes, rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If skin/eye irritation occurs, get medical advice/attention.

DOTInformation : Nonhazardous

Recommended Storage : Ambient