missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Manganese Standard For ICP, 10,000μg/mL in 5% HNO3, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

$292.23 - $550.40

Chemical Identifiers

For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Chemical Name or Material Manganese Standard
View More Specs

Disclaimers:

Product intended for laboratory use only


Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
PLMN2-3Y SDS Spex Certiprep Llc
PLMN23Y
Triple-leached LDPE bottle 125mL Each for $292.23
PLMN2-3X SDS Spex Certiprep Llc
PLMN23X
Triple-leached LDPE bottle 500mL Each for $550.40
Specifications

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Mn10,000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Chemical Identifiers

AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Manganese Standard

Specifications

125mL
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
7732-18-5,7439-96-5,7697-37-2
Assurance Grade
Triple-leached LDPE bottle
5% HNO3
Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
Manganese Standard

Safety and Handling

DOTInformation : Hazardous

Recommended Storage : Ambient