Please login to your online account to display your discounted pricing

Manganese Standard, 1000μg/mL in water with dilute nitric acid, ICP-025, Ultra Scientific


Quantity 125mL

Manufacturer: ultra scientific  ICP025

 View more versions of this product

Catalog No. US-ICP-025

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 125mL
Analyte or Component Names Water,Nitric acid,Manganese
Solvent or Matrix Water with dilute nitric acid
CAS Max % 0.1
Chemical Name or Material Manganese Standard
CAS 007732-18-5
CAS Max % 97.9
CAS 007697-37-2
CAS Max % 2.0
Concentration or Composition (by Analyte or Components) 1000μg/mL
CAS 007439-96-5

Specifications

Chemical Name or Material Manganese Standard
CAS 007732-18-5
CAS Max % 97.9
CAS 007697-37-2
CAS Max % 2.0
CAS 007439-96-5
CAS Max % 0.1