Please login to your online account to display your discounted pricing

Manganese Standard, 1000μg/mL in water with dilute nitric acid, ICP-025, Ultra Scientific

$94.74 - $94.74

Chemical Identifiers

Chemical Name or Material Manganese Standard
CAS 007732-18-5
CAS Max % 97.9
CAS 007697-37-2
CAS Max % 2.0
CAS 007439-96-5
CAS Max % 0.1
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

US-ICP-025

 
ultra scientific
ICP025
125mL Each for $94.74
Description & Specifications

Specifications

Quantity 125mL
Solvent or Matrix Water with dilute nitric acid
Analyte or Component Names Water,Nitric acid,Manganese
CAS Max % 0.1
Chemical Name or Material Manganese Standard
CAS 007697-37-2
CAS 007732-18-5
CAS Max % 2.0
CAS Max % 97.9
CAS 007439-96-5
Concentration or Composition (by Analyte or Components) 1000μg/mL