Please login to your online account to display your discounted pricing

Alfa Aesar™ Manganese sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.95% (metals basis)

$1,868.38 - $1,868.38

Chemical Identifiers

Chemical Name or Material Manganese sputtering target
CAS 7439-96-5
Name Note 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
MDL Number MFCD00011111
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40865KS

  SDS
alfa aesar™
40865KS
1 each Each for $1,868.38
Description & Specifications

Specifications

Physical Form Hot Pressed
Chemical Name or Material Manganese sputtering target
TSCA TSCA
Name Note 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
Assay 99.95%
Assay Percent Notes (metals basis)
MDL Number MFCD00011111
CAS 7439-96-5
Odor Odorless
Quantity 1 each