Please login to your online account to display your discounted pricing

Alfa Aesar™ Manganese sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.95% (metals basis)

$1,378.69 - $1,378.69

Chemical Identifiers

Chemical Name or Material Manganese sputtering target
CAS 7439-96-5
Name Note 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
MDL Number MFCD00011111
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40864KS

  SDS
alfa aesar™
40864KS
1 each Each for $1,378.69
Description & Specifications

Specifications

Physical Form Hot Pressed
TSCA TSCA
Assay Percent Notes (metals basis)
MDL Number MFCD00011111
Quantity 1 each
Assay 99.95%
Odor Odorless
Name Note 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
CAS 7439-96-5
Chemical Name or Material Manganese sputtering target