Please login to your online account to display your discounted pricing

Alfa Aesar™ Manganese sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.95% (metals basis)

$1,350.05 - $1,350.05

Chemical Identifiers

Chemical Name or Material Manganese sputtering target
CAS 7439-96-5
Name Note 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
MDL Number MFCD00011111
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40863KS

  SDS
alfa aesar™
40863KS
1 each Each for $1,350.05
Description & Specifications

Specifications

Physical Form Hot Pressed
Name Note 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
CAS 7439-96-5
Assay Percent Notes (metals basis)
TSCA TSCA
MDL Number MFCD00011111
Quantity 1 each
Assay 99.95%
Chemical Name or Material Manganese sputtering target
Odor Odorless