Please login to your online account to display your discounted pricing

Manganese, Reference Standard Solution (1000ppm ± 1%/Certified), Fisher Chemical


Quantity 500mL
Packaging Poly Bottle

Manufacturer: fisher scientific company  SM81500

 View more versions of this product

Catalog No. SM81-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Boiling Point 100°C
Purity Grade Certified Grade
Quantity 500mL
DOT Information DOT Class 8, : Corrosive
Physical Form Liquid
Color Colorless
Vapor Pressure 14mmHg at 20°C
Concentration or Composition (by Analyte or Components) 1000ppm ±1%
Packaging Poly Bottle
MDL Number MFCD00011111
Traceability to NIST LOT NUMBER USED Manganese ref. Std.
ChemAlert Storage Symbol White
Concentration 990 to 1010ppm
CAS 7732-18-5
CAS 17141-63-8
Chemical Name or Material Manganese, Reference Standard Solution
Name Note 1000ppm ± 1%
CAS 7697-37-2

Specifications

Chemical Name or Material Manganese, Reference Standard Solution
Name Note 1000ppm ± 1%
CAS 17141-63-8
CAS 7697-37-2
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00011111
Accessories
 

SDS & Safety Documents