Please login to your online account to display your discounted pricing

Manganese, Reference Standard Solution (1000ppm ± 1%/Certified), Fisher Chemical

$78.59 - $92.82

Chemical Identifiers

Chemical Name or Material Manganese, Reference Standard Solution
Name Note 1000ppm ± 1%
CAS 17141-63-8
CAS 7697-37-2
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00011111
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SM81-500

  SDS
fisher scientific company
SM81500
Poly Bottle 500mL Each for $92.82
Case of 6 Each for $556.92

SM81-100

  SDS
fisher scientific company
FLSM81100
Poly Bottle 100mL Each for $78.59
Case of 6 Each for $471.54
Description & Specifications

Specifications

Boiling Point 100°C
Purity Grade Certified Grade
Quantity 500mL
DOT Information DOT Class 8, : Corrosive
Physical Form Liquid
Color Colorless
Vapor Pressure 14mmHg at 20°C
Concentration or Composition (by Analyte or Components) 1000ppm ±1%
Packaging Poly Bottle
MDL Number MFCD00011111
Traceability to NIST LOT NUMBER USED Manganese ref. Std.
ChemAlert Storage Symbol White
Concentration 990 to 1010ppm
CAS 7732-18-5
CAS 17141-63-8
Chemical Name or Material Manganese, Reference Standard Solution
Name Note 1000ppm ± 1%
CAS 7697-37-2
Safety and Handling