missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Manganese Powder, -325 mesh, 99.3% (Metals basis), Alfa Aesar™

Form: Apparent density 2.6-3.5g/cm3

Manufacturer:  Thermo Fisher Scientific 1023814

 View more versions of this product

Catalog No. AA1023814



Specifications

Specifications

Manganese
7439-96-5
99.3% (Metals basis)
MFCD00011111
colloidal, compounds, cutaval, fume, magnacat, mangan, mangan polish, manganese, elemental, metal alloy, tronamang
[Mn]
54.938
CHEBI:35154
Gray
25g
Gray powder
Mn
-325 Mesh
PWHULOQIROXLJO-UHFFFAOYSA-N
manganese
23930
Powder, Apparent density 2.6-3.5g/cm

Chemical Identifiers

7439-96-5
54.938
PWHULOQIROXLJO-UHFFFAOYSA-N
23930
manganese
Mn
MFCD00011111
colloidal, compounds, cutaval, fume, magnacat, mangan, mangan polish, manganese, elemental, metal alloy, tronamang
CHEBI:35154
[Mn]
Safety and Handling

Safety and Handling

P210-P240-P241-P264b-P280-P305+P351+P338-P370+P378q

H228-H319

DOTInformation : DOT Class: 4.1, Packaging Group: III

EINECSNumber : 231-105-1

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents