missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Manganese Powder, <10 micron, 99.6% (Metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 4509818

 View more versions of this product

Catalog No. AA4509818



Specifications

Specifications

Manganese
7439-96-5
99.6% (Metals basis)
MFCD00011111
PWHULOQIROXLJO-UHFFFAOYSA-N
manganese
23930
Powder
50g
Gray powder
Mn
colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds
[Mn]
54.938
CHEBI:35154
Gray

Chemical Identifiers

7439-96-5
54.938
PWHULOQIROXLJO-UHFFFAOYSA-N
23930
manganese
Mn
MFCD00011111
colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds
CHEBI:35154
[Mn]
Safety and Handling

Safety and Handling

Take precautionary measures against static discharges.

H228: Flammable solid.

DOTInformation : DOT Class: 4.1, Packaging Group: III

EINECSNumber : 231-105-1

SDS Documents