missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Manganese plasma standard solution, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 13826AP

 View more versions of this product

Catalog No. AA13826AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Manganese
Clear liquid
MFCD00011111
PWHULOQIROXLJO-UHFFFAOYSA-N
manganese
23930
Liquid
Colorless
7439-96-5,7697-37-2,7732-18-5
Mn
Nitric acid: 5.0%; Manganese: 0.1%; Water: 94.9%
[Mn]
54.938
CHEBI:35154
23 hPa (17mm Hg) at 20°C
500mL

Chemical Identifiers

7439-96-5
54.938
PWHULOQIROXLJO-UHFFFAOYSA-N
CHEBI:35154
[Mn]
Mn
MFCD00011111
23930
manganese
Safety and Handling

Safety and Handling

H314: Causes severe skin burns and eye damage.

DOTInformation : Hazard Class: 8, Packing Group: III

EINECSNumber : 231-714-2

Documents