missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Manganese pieces, irregular, 99.9% (metals basis), Alfa Aesar

Manufacturer:  Thermo Fisher Scientific 10236A7

 View more versions of this product

Catalog No. AA10236A7



Specifications

Specifications

Manganese
7439-96-5
99.9% (Metals basis)
MFCD00011111
PWHULOQIROXLJO-UHFFFAOYSA-N
manganese
23930
54.94
Gray
5kg
Gray irregularly shaped pieces
Mn
colloidal
[Mn]
54.938
CHEBI:35154
Irregular Shaped Pieces

Chemical Identifiers

7439-96-5
54.938
PWHULOQIROXLJO-UHFFFAOYSA-N
23930
manganese
Mn
MFCD00011111
colloidal
CHEBI:35154
[Mn]
Safety and Handling

Safety and Handling

EINECSNumber : 231-105-1

Documents