missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Manganese phosphide, 99% (metals basis)

Manufacturer:  Alfa Aesar™ 1402004

 View more versions of this product

Catalog No. AA1402004



Specifications

Specifications

Manganese phosphide
(metals basis)
226.75
99%
Odorless
2g
12263-33-1
MFCD00064736
Powder
−100 Mesh
1095°C

Chemical Identifiers

12263-33-1
MFCD00064736
Documents