Please login to your online account to display your discounted pricing

Alfa Aesar™ Manganese, Oil based standard solution, Specpure™, Mn 1000μg/g

Chemical Identifiers

Chemical Name or Material Manganese Oil based standard solution
Name Note Mn 1000μg/g
Purity Grade Specpure™
MDL Number MFCD00011111
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1421418

 
alfa aesar™
1421418
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Manganese Oil based standard solution
Quantity 50g
Name Note Mn 1000μg/g
Purity Grade Specpure™
Physical Form Liquid
MDL Number MFCD00011111