Please login to your online account to display your discounted pricing

Alfa Aesar™ Manganese(IV) telluride, 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Manganese(IV) telluride
CAS 12032-89-2
Formula Weight 310.14
MDL Number MFCD00064703
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1313418

  SDS
alfa aesar™
1313418
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1313409

  SDS
alfa aesar™
1313409
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 310.14
Chemical Name or Material Manganese(IV) telluride
TSCA TSCA
Odor Odorless
Particle Size 6mm & down
Physical Form Pieces
Assay 99.9%
Assay Percent Notes (metals basis)
MDL Number MFCD00064703
CAS 12032-89-2
Quantity 50g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.