missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Manganese(IV) telluride, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 1313418

 View more versions of this product

Catalog No. AA1313418



Specifications

Specifications

Manganese(IV) telluride
(metals basis)
MFCD00064703
KDJKBQYOEWVJGP-UHFFFAOYSA-N
bis(tellanylidene)manganese
82829
Pieces
Odorless
50g
12032-89-2
MnTe2
manganese ditelluride, manganese iv telluride, bis tellanylidene manganese, ditellanylidenemanganese, manganese telluride
[Mn](=[Te])=[Te]
310.138
310.14
99.9%
6mm & down

Chemical Identifiers

12032-89-2
310.138
KDJKBQYOEWVJGP-UHFFFAOYSA-N
82829
[Mn](=[Te])=[Te]
MnTe2
MFCD00064703
manganese ditelluride, manganese iv telluride, bis tellanylidene manganese, ditellanylidenemanganese, manganese telluride
bis(tellanylidene)manganese
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Documents