Learn More
Manganese(IV) oxide, 99.99+%, (trace metal basis), Thermo Scientific™
Supplier: Thermo Scientific Chemicals 193471000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
For ultra trace analysis, Oxidation reagentSpecifications
Manganese(IV) oxide | |
100g | |
MnO2 | |
MFCD00003463 | |
15,5795 | |
Solubility in water: insoluble. Other solubilities: soluble in hydrochloric acid | |
O=[Mn]=O | |
86.94 | |
86.94 | |
Trace Metal Basis |
1313-13-9 | |
99.99+% | |
MnO2 | |
01,637; 02,257; 03,191; 04,317; 05,422; 06,357; 08,312; 13,233; 14,200; 15,197; 16,202 | |
manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein | |
NUJOXMJBOLGQSY-UHFFFAOYSA-N | |
dioxomanganese | |
14801 | |
99.99+% | |
Glass bottle |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure if inhaled.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
D
GHS Signal Word: Warning
EINECSNumber : 215-202-6
RUO â Research Use Only