missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Manganese(III) oxide, 98%, Alfa Aesar™

Manufacturer:  Alfa Aesar 87791A3

 View more versions of this product

Catalog No. AA87791A3



Specifications

Specifications

Manganese (III) oxide
98%
MFCD00016217
manganese iii oxide, manganic oxide, dimanganese trioxide, manganese sesquioxide, manganese trioxide, manganese manganate, manganese sisquioxide, manganese 3+ oxide, manganese oxide, oxo oxomanganiooxy manganese
O=[Mn]O[Mn]=O
157.873
157.86
98%
−325 Mesh
Black
2kg
1317-34-6
Mn2O3
14,5737
GEYXPJBPASPPLI-UHFFFAOYSA-N
oxo(oxomanganiooxy)manganese
14824
Powder
4.50g/mL
Odorless
1080°C (decomposition)

Chemical Identifiers

1317-34-6
157.873
GEYXPJBPASPPLI-UHFFFAOYSA-N
14824
O=[Mn]O[Mn]=O
Mn2O3
MFCD00016217
manganese iii oxide, manganic oxide, dimanganese trioxide, manganese sesquioxide, manganese trioxide, manganese manganate, manganese sisquioxide, manganese 3+ oxide, manganese oxide, oxo oxomanganiooxy manganese
oxo(oxomanganiooxy)manganese
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332-H315-H319-H335
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : OP0915000

TSCA : TSCA

SDS Documents