missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Manganese(III) fluoride, 98% (metals basis)

Manufacturer:  Alfa Aesar™ 1306918

 View more versions of this product

Catalog No. AA1306918



Specifications

Specifications

Manganese(III) fluoride
(metals basis)
UN3288
Insoluble in organic solvents. Hydrolyzed by water.
Crystalline
3.54 g/mL
(decomposition)
7783-53-1
MFCD00016223
14,5741
111.93
98%
Hygroscopic
50g

Chemical Identifiers

7783-53-1
MFCD00016223
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P220-P221-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c

H272-H302+H332-H315-H318-H335

DOTInformation : Hazard Class: 5.1; Packaging Group: II

EINECSNumber : 232-006-6

RTECSNumber : OP0882600

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents