missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Manganese(III) acetate meso-tetraphenylporphine

$58.81 - $170.86

Chemical Identifiers

CAS 58356-65-3
Formula Weight 726.71
MDL Number MFCD00216548
Chemical Name or Material Manganese(III) acetate meso-tetraphenylporphine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3930803 SDS Alfa Aesar™
3930803
1g Each for $170.86
AA39308MD SDS Alfa Aesar™
39308MD
250mg Each for $58.81
Specifications Description & Specifications

Chemical Identifiers

58356-65-3
726.71
MFCD00216548
Manganese(III) acetate meso-tetraphenylporphine

Specifications

1g
Powder
MFCD00216548
58356-65-3
726.71
Manganese(III) acetate meso-tetraphenylporphine

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning