Please login to your online account to display your discounted pricing

Manganese(III) acetate dihydrate, 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: fisher scientific acros  201741000

 View more versions of this product

Catalog No. AC201741000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Powder
Formula Weight 268.09
Quantity 100g
Color Orange-brown to brown
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Suspected of damaging fertility.
Chemical Name or Material Manganese(III) acetate dihydrate
Name Note 98%
Molecular Formula C6H9MnO6·2H2O
Linear Formula (CH3CO2)3Mn·2H2O
CAS 19513-05-4
Assay Percent Range 98%
MDL Number MFCD00150022
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Manganese(III) acetate dihydrate
Name Note 98%
CAS 19513-05-4
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H9MnO6·2H2O
Linear Formula (CH3CO2)3Mn·2H2O
Formula Weight 268.09
MDL Number MFCD00150022
Accessories

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Suspected of damaging fertility.

SDS & Safety Documents