missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Manganese(III) 2,4-pentanedionate

Manufacturer:  Alfa Aesar™ 1253009

 View more versions of this product

Catalog No. AA1253009



Specifications

Specifications

Manganese(III) 2,4-pentanedionate
~160°C (decomposition)
14284-89-0
MFCD00000023
Soluble in benzene, ethyl acetate
CC(=CC(=O)C)[O-].[Mn+3]
154.047
352.24
Hygroscopic
10g
C5H7MnO2+2
mn iii acetylacetonate
BCOBLSSFEZDWKH-BJILWQEISA-M
manganese(3+);(E)-4-oxopent-2-en-2-olate
122130696
Powder

Chemical Identifiers

14284-89-0
154.047
BCOBLSSFEZDWKH-BJILWQEISA-M
122130696
CC(=CC(=O)C)[O-].[Mn+3]
C5H7MnO2+2
MFCD00000023
mn iii acetylacetonate
manganese(3+);(E)-4-oxopent-2-en-2-olate
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 238-188-3

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents