Please login to your online account to display your discounted pricing

Manganese(II) titanium oxide, Yellow powder, 99.9% (Metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Manganese(II) titanium oxide
CAS 12032-74-5
Physical Form Powder
Assay Percent Range 99.9% (Metals basis)
Molecular Formula MnTiO3
Formula Weight 150.84
MDL Number MFCD00016216
Synonym Manganese titanate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1313309

  SDS
alfa aesar
1313309
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1313318

  SDS
alfa aesar
1313318
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 150.84
Quantity 10g
Color Yellow
Density 4.54g/cm3 at 20°C
Chemical Name or Material Manganese(II) titanium oxide
Odor Odorless
Assay Percent Range 99.9% (Metals basis)
Molecular Formula MnTiO3
MDL Number MFCD00016216
CAS 12032-74-5
Synonym Manganese titanate