missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Manganese (II) Sulfate Solution (APHA), Fisher Chemical

For determination of dissolved oxygen

Manufacturer:  Fisher Scientific Company SM20500

 View more versions of this product

Catalog No. SM20-500



 

Specifications Description & Specifications

Specifications

Manganese (II) Sulfate Solution
7785-87-7,7732-18-5
Glass Bottle
Liquid
Light Red
500mL
Conforms to the standards of APHA
Pass Test
1.26 to 1.28 (at 25°C)
1.27g/cm³
MFCD00149159

Chemical Identifiers

7785-87-7,7732-18-5
MFCD00149159
Manganese (II) Sulfate Solution, Conforms to the standards of APHA

Safety and Handling

SDS & Safety Documents