Please login to your online account to display your discounted pricing

Manganese (II) Sulfate Solution (APHA), Fisher Chemical


For determination of dissolved oxygen

Quantity 1L
Packaging Glass Bottle

Manufacturer: fisher scientific company  SM201

 View more versions of this product

Catalog No. SM20-1

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Quantity 1L
Physical Form Liquid
Color Light Red
Density 1.27g/cm³
Packaging Glass Bottle
Specific Gravity 1.26 to 1.28 (at 25°C)
CAS 7732-18-5
ChemAlert Storage Symbol Gray
MDL Number MFCD00149159
Identification Pass Test
CAS 7785-87-7
Name Note Conforms to the standards of APHA
Chemical Name or Material Manganese (II) Sulfate Solution

Specifications

Chemical Name or Material Manganese (II) Sulfate Solution
Name Note Conforms to the standards of APHA
CAS 7785-87-7
CAS 7732-18-5
MDL Number MFCD00149159
Accessories
 

SDS & Safety Documents