Please login to your online account to display your discounted pricing

Manganese (II) Sulfate Solution (APHA), Fisher Chemical

For determination of dissolved oxygen

$242.54 - $291.23

Chemical Identifiers

Chemical Name or Material Manganese (II) Sulfate Solution
Name Note Conforms to the standards of APHA
CAS 7785-87-7
CAS 7732-18-5
MDL Number MFCD00149159
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SM20-1

  SDS
fisher scientific company
SM201
Glass Bottle 1L Each for $291.23
Case of 6 Each for $1,747.38

SM20-500

  SDS
fisher scientific company
SM20500
Glass Bottle 500mL Each for $242.54
Case of 6 Each for $1,455.24
Description & Specifications

Specifications

Quantity 1L
Physical Form Liquid
Color Light Red
Density 1.27g/cm³
Packaging Glass Bottle
Specific Gravity 1.26 to 1.28 (at 25°C)
CAS 7732-18-5
ChemAlert Storage Symbol Gray
MDL Number MFCD00149159
Identification Pass Test
CAS 7785-87-7
Name Note Conforms to the standards of APHA
Chemical Name or Material Manganese (II) Sulfate Solution

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.

Safety and Handling