missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell Fluka™ Manganese(II) sulfate monohydrate, Puriss., meets analytical spec. of Ph. Eur, BP, USP, FCC, 99 to 100.5% (calc. for dried substance), Honeywell Fluka™

Puriss., meets analytical specification of Ph. Eur, BP, USP, FCC, 99-100.5% (calc. for dried substance)

Manufacturer:  Honeywell Fluka™ 132451KG

 View more versions of this product

Catalog No. 6000919



Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Specifications

Manganese(Ii) Sulfate Monohydrate
10034-96-5
MnSO4 · H2O
3077
ISPYRSDWRDQNSW-UHFFFAOYSA-L
manganese(2+);sulfate;hydrate
177577
169.02g/mol
puriss.
Meets analytical specification of Ph. Eur.BP, USP, FCC
H2MnO5S
MFCD00149159
manganese 2+ sulfate hydrate
O.[O-]S(=O)(=O)[O-].[Mn+2]
169.009
CHEBI:86364
99 to 100.5% (calc. for dried substance)
1kg

Chemical Identifiers

10034-96-5
169.009
ISPYRSDWRDQNSW-UHFFFAOYSA-L
177577
manganese(2+);sulfate;hydrate
H2MnO5S
MFCD00149159
manganese 2+ sulfate hydrate
CHEBI:86364
O.[O-]S(=O)(=O)[O-].[Mn+2]
Safety and Handling

Safety and Handling

P260

H373-H411

EINECSNumber : 232-089-9

RTECSNumber : OP0893500

Documents