Learn More
Manganese(II) sulfate monohydrate, 98+%, ACS reagent, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 423915000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
850.0°C | |
700.0°C | |
Pink | |
500 g | |
H2MnO5S | |
MFCD00149159 | |
manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate | |
ISPYRSDWRDQNSW-UHFFFAOYSA-L | |
manganese(2+);sulfate;hydrate | |
177577 | |
169.02 | |
ACS Reagent | |
Plastic bottle |
Manganese(II) sulfate monohydrate | |
10034-96-5 | |
Powder | |
98.0 to 101% (Complexometry) | |
MnSO4·H2O | |
15, 5804 | |
Solubility in water: soluble. Other solubilities: insoluble in alcohol | |
O.[Mn++].[O-]S([O-])(=O)=O | |
169.01 | |
CHEBI:86364 | |
≥98% | |
10.0% to 12.0% |
Chemical Identifiers
10034-96-5 | |
169.01 | |
ISPYRSDWRDQNSW-UHFFFAOYSA-L | |
177577 | |
O.[Mn++].[O-]S([O-])(=O)=O |
H2MnO5S | |
MFCD00149159 | |
manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate | |
CHEBI:86364 |
Safety and Handling
GHS H Statement
May cause damage to organs through prolonged or repeated exposure if inhaled.
Causes serious eye damage.
Toxic to aquatic life with long lasting effects.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately
GHS Signal Word: Danger
RTECSNumber : OP0893500
RUO – Research Use Only