Please login to your online account to display your discounted pricing

Alfa Aesar™ Manganese(II) selenide, 99.9%

Chemical Identifiers

Chemical Name or Material Manganese(II) selenide
CAS 1313-22-0
Molecular Formula MnSe
Formula Weight 133.90g/mol
MDL Number MFCD00049486
UN Number UN3283
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1708903

 
alfa aesar™
A17089-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material Manganese(II) selenide
CAS 1313-22-0
Assay 99.9%
Molecular Formula MnSe
Formula Weight 133.90g/mol
MDL Number MFCD00049486
UN Number UN3283
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3 GHS P Statement
P260-P261-P301+P310-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S.
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P260-P261-P301+P310-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.