missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Manganese(II) perchlorate hexahydrate, 99.995% (metals basis)

Manufacturer:  Alfa Aesar™ 4431830

 View more versions of this product

Catalog No. AA4431830



Specifications

Specifications

Manganese (II) perchlorate hexahydrate
(metals basis)
MFCD00150255
2clo4.mn.6h2o
AIFGVOMWPFMOCN-UHFFFAOYSA-L
manganese(2+);diperchlorate;hexahydrate
203855
Crystalline
Odorless
165°C (decomposition)
15364-94-0
Cl2H12MnO14
UN1481
Soluble in water,benzene,ethyl acetate
O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Mn+2]
361.92
361.93 (253.84 Anhydrous)
99.995%
Hygroscopic
250g

Chemical Identifiers

15364-94-0
361.92
AIFGVOMWPFMOCN-UHFFFAOYSA-L
203855
O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Mn+2]
Cl2H12MnO14
MFCD00150255
2clo4.mn.6h2o
manganese(2+);diperchlorate;hexahydrate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P220-P221-P280-P370+P378q-P501c

H272

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.

EINECSNumber : 237-390-9

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Documents