Learn More
Manganese(II) oxalate dihydrate, Mn 30% min, Thermo Scientific Chemicals
As a paint and varnish drier
Supplier: Thermo Scientific Chemicals 01468530
Description
Manganese(II) oxalate dihydrate is used as a paint and varnish drier. It is also used as a chemical reagent as well as to prepare other manganese compounds. In addition, it is used as a semiconductor photosensitive material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Manganese(II) oxalate dihydrate | |
100°C (decomposition) | |
2.453 g/mL | |
C2H4MnO6 | |
250 g | |
manganese ii oxalate 2-hydrate | |
HDJUVFZHZGPHCQ-UHFFFAOYSA-L | |
manganese(2+);oxalate;dihydrate | |
131882857 | |
Mn 30% min. |
6556-16-7 | |
White | |
Odorless | |
MFCD00150452 | |
14,5734 | |
Soluble in dilute acids; Slightly soluble in water. | |
C(=O)(C(=O)[O-])[O-].O.O.[Mn+2] | |
178.986 | |
178.99 (142.96 Anhydrous) | |
Chunks and/or Powder |
Chemical Identifiers
6556-16-7 | |
178.986 | |
HDJUVFZHZGPHCQ-UHFFFAOYSA-L | |
131882857 | |
C(=O)(C(=O)[O-])[O-].O.O.[Mn+2] |
C2H4MnO6 | |
MFCD00150452 | |
manganese ii oxalate 2-hydrate | |
manganese(2+);oxalate;dihydrate |
Safety and Handling
GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
H302+H312
EINECSNumber : 211-367-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only